1-(4-Bromphenyl)-4-(ethylsulfonyl)piperazin
Lieferant: COMBI-BLOCKS
AM-2076-5G
AM-2076-1G
COBBAM-2076-5GEA
884
EUR
COBBAM-2076-5G
COBBAM-2076-1G
1-(4-Bromphenyl)-4-(ethylsulfonyl)piperazin
1-(4-Bromphenyl)-4-(ethylsulfonyl)piperazin
Formel:
C₁₂H₁₇BrN₂O₂S |
MDL:
MFCD16036099 CAS-Nummer: 1221658-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...