1-(4-Bromphenyl)-4-(methylsulfonyl)piperazin
Lieferant: COMBI-BLOCKS
AM-2075-5G
AM-2075-1G
COBBAM-2075-5GEA
884
EUR
COBBAM-2075-5G
COBBAM-2075-1G
1-(4-Bromphenyl)-4-(methylsulfonyl)piperazin
1-(4-Bromphenyl)-4-(methylsulfonyl)piperazin
Formel:
C₁₁H₁₅BrN₂O₂S |
MDL:
MFCD14584697 CAS-Nummer: 357647-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...