1H-Benzotriazol-1-methanol

Lieferant: CHEMPUR

Synonyme: 1-(Hydroxymethyl)benzotriazole

XBX00334.250 XBX00334.1
CHMPXBX00334.250EA 0 EUR
CHMPXBX00334.250 MAYBXBX00334.1
1H-Benzotriazol-1-methanol
1H-Benzotriazol-1-methanol
Formel: C₇H₇N₃O
Molecular Weight: 149,1500 g/mol
MDL: MFCD00179118
CAS-Nummer: 28539-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR