1H-Benzotriazol-1-methanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(Hydroxymethyl)benzotriazole
M-Clarity™ Qualitätsstufe = MQ100
410233-10G
410233-50G
SIAL410233-10GEA
48.8
EUR
SIAL410233-10G
SIAL410233-50G
1H-Benzotriazol-1-methanol, Sigma-Aldrich®
1H-Benzotriazol-1-methanol
Formel:
C₇H₇N₃O Molecular Weight: 149,1500 g/mol |
MDL:
MFCD00179118 CAS-Nummer: 28539-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...