2-(Ethoxycarbonyl)furan-5-boronsäurepinakolester 96%

Lieferant: Thermo Fisher Scientific

Synonyme: Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate

H52312.MD H52312.03
H52312.MDEA 84.3 EUR
H52312.MD H52312.03
2-(Ethoxycarbonyl)furan-5-boronsäurepinakolester 96%
2-(Ethoxycarbonyl)furan-5-boronsäurepinakolester

Purity: 96% CAS No.: 1073338-92-7 Molecular Formula: C13H19BO5 Formula Weight: 266.10

Formel: C₁₃H₁₉BO₅
Molecular Weight: 266.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11855980
CAS-Nummer: 1073338-92-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR