3-(Methoxycarbonyl)furan-2-boronsäure

Lieferant: COMBI-BLOCKS

BB-5781-1G BB-5781-250M
COBBBB-5781-1GEA 1240 EUR
COBBBB-5781-1G COBBBB-5781-250M
3-(Methoxycarbonyl)furan-2-boronsäure
3-(Methoxycarbonyl)furan-2-boronsäure
Formel: C₆H₇BO₅
MDL: MFCD12025980
CAS-Nummer: 868286-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR