2-(Ethoxycarbonyl)furan-5-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-(Ethoxycarbonyl)furan-2-yl)boronic acid

BB-5514-1G BB-5514-250M
COBBBB-5514-1GEA 449 EUR
COBBBB-5514-1G COBBBB-5514-250M
2-(Ethoxycarbonyl)furan-5-boronsäure
2-(Ethoxycarbonyl)furan-5-boronsäure
Formel: C₇H₉BO₅
Molecular Weight: 183,9544 g/mol
MDL: MFCD10696632
CAS-Nummer: 1150114-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR