(2-Phenoxyethyl)triphenylphosphoniumbromid
Lieferant: Thermo Scientific
JFD04038.250
JFD04038.1
MAYBJFD04038.250EA
0
EUR
MAYBJFD04038.250
MAYBJFD04038.1
(2-Phenoxyethyl)triphenylphosphoniumbromid
(2-Phenoxyethyl)triphenylphosphonium bromide
Formel:
C₂₆H₂₄BrOP |
CAS-Nummer:
22409-83-2 EINECS: 244-963-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...