Tetrakis(hydroxymethyl)phosphoniumchlorid ≥80%, Sigma-Aldrich®

Lieferant: Merck

404861-100ML 404861-500ML
SIAL404861-100MLEA 61.6 EUR
SIAL404861-100ML SIAL404861-500ML
Tetrakis(hydroxymethyl)phosphoniumchlorid ≥80%, Sigma-Aldrich®
Tetrakis(hydroxymethyl)phosphoniumchlorid
Formel: C₄H₁₂ClO₄P
Molecular Weight: 190,56 g/mol
MDL: MFCD00031687
CAS-Nummer: 124-64-1
EINECS: 204-707-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR