1-[(Ammoniooxy)methyl]-2-brombenzolchlorid
Lieferant: BIONET RESEARCH
1G-915.500
1G-915.1
1G-915.5
1G-915.10
BIOT1G-915.500EA
0
EUR
BIOT1G-915.500
BIOT1G-915.1
BIOT1G-915.5
BIOT1G-915.10
1-[(Ammoniooxy)methyl]-2-brombenzolchlorid
1-[(Ammoniooxy)methyl]-2-brombenzolchlorid
Formel:
C₇H₉BrClNO |
MDL:
MFCD01114573 CAS-Nummer: 51572-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...