1-[(Ammoniooxy)methyl]-4-(methoxycarbonyl)benzolchlorid

Lieferant: BIONET RESEARCH

7G-944.500 7G-944.1 7G-944.5 7G-944.10
BIOT7G-944.500EA 0 EUR
BIOT7G-944.500 BIOT7G-944.1 BIOT7G-944.5 BIOT7G-944.10
1-[(Ammoniooxy)methyl]-4-(methoxycarbonyl)benzolchlorid
1-[(Ammoniooxy)methyl]-4-(methoxycarbonyl)benzolchlorid
Formel: C₉H₁₂ClNO₃
MDL: MFCD01114498
CAS-Nummer: 78254-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR