1-[(Ammoniooxy)methyl]-2-(trifluormethyl)benzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-[2-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride

1G-933.500 1G-933.1 1G-933.5 1G-933.10
BIOT1G-933.500EA 0 EUR
BIOT1G-933.500 BIOT1G-933.1 BIOT1G-933.5 BIOT1G-933.10
1-[(Ammoniooxy)methyl]-2-(trifluormethyl)benzolchlorid
1-[(Ammoniooxy)methyl]-2-(trifluormethyl)benzolchlorid
Formel: C₈H₉ClF₃NO
MDL: MFCD01114578
CAS-Nummer: 215599-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR