1-[(Ammoniooxy)methyl]-2-chlor-4-fluorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(2-Chloro-4-fluorobenzyl)hydroxylamine hydrochloride

1G-925.500 1G-925.1 1G-925.5 1G-925.10
BIOT1G-925.500EA 0 EUR
BIOT1G-925.500 BIOT1G-925.1 BIOT1G-925.5 BIOT1G-925.10
1-[(Ammoniooxy)methyl]-2-chlor-4-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-2-chlor-4-fluorbenzolchlorid
Formel: C₇H₈Cl₂FNO
MDL: MFCD01114574
CAS-Nummer: 317821-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR