2-[(Ammoniooxy)methyl]-1-chlor-3-fluorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(2-Chloro-6-fluorobenzyl)hydroxylamine hydrochloride

12E-928.500 12E-928.1 12E-928.5 12E-928.10
BIOT12E-928.500EA 0 EUR
BIOT12E-928.500 BIOT12E-928.1 BIOT12E-928.5 BIOT12E-928.10
2-[(Ammoniooxy)methyl]-1-chlor-3-fluorbenzolchlorid
2-[(Ammoniooxy)methyl]-1-chlor-3-fluorbenzolchlorid
Formel: C₇H₈Cl₂FNO
MDL: MFCD01114572
CAS-Nummer: 93081-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR