1-[(Ammoniooxy)methyl]-2-fluorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(2-Fluorobenzyl)hydroxylamine hydrochloride

4F-951.500 4F-951.1 4F-951.5 4F-951.10
BIOT4F-951.500EA 0 EUR
BIOT4F-951.500 BIOT4F-951.1 BIOT4F-951.5 BIOT4F-951.10
1-[(Ammoniooxy)methyl]-2-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-2-fluorbenzolchlorid
Formel: C₇H₉ClFNO
Molecular Weight: 177.6039 g/mol
MDL: MFCD01114584
CAS-Nummer: 215599-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR