1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-[3-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride

12E-926.500 12E-926.1 12E-926.5 12E-926.10
BIOT12E-926.500EA 0 EUR
BIOT12E-926.500 BIOT12E-926.1 BIOT12E-926.5 BIOT12E-926.10
1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
Formel: C₈H₉ClF₃NO
MDL: MFCD01114571
CAS-Nummer: 15256-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR