1-[(Ammoniooxy)methyl]-3-fluorbenzolchlorid
Lieferant: BIONET RESEARCH
Synonyme:
O-(3-Fluorobenzyl)hydroxylamine hydrochloride
4F-952.500
4F-952.1
4F-952.5
4F-952.10
BIOT4F-952.500EA
0
EUR
BIOT4F-952.500
BIOT4F-952.1
BIOT4F-952.5
BIOT4F-952.10
1-[(Ammoniooxy)methyl]-3-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-3-fluorbenzolchlorid
Formel:
C₇H₉ClFNO |
MDL:
MFCD01114585 CAS-Nummer: 51572-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...