1-[(Ammoniooxy)methyl]-3-fluorbenzolchlorid

Lieferant: FLUOROCHEM

Synonyme: O-(3-Fluorobenzyl)hydroxylamine hydrochloride

010010-1G 010010-5G
FLUO010010-1GEA 479 EUR
FLUO010010-1G FLUO010010-5G
1-[(Ammoniooxy)methyl]-3-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-3-fluorbenzolchlorid
Formel: C₇H₉ClFNO
MDL: MFCD01114585
CAS-Nummer: 51572-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR