1-[(Ammoniooxy)methyl]-4-(trifluormethyl)benzolchlorid
Lieferant: BIONET RESEARCH
Synonyme:
O-(4-(Trifluoromethyl)benzyl)hydroxylamine hydrochloride
8H-028.500
8H-028.1
8H-028.5
8H-028.10
BIOT8H-028.500EA
0
EUR
BIOT8H-028.500
BIOT8H-028.1
BIOT8H-028.5
BIOT8H-028.10
1-[(Ammoniooxy)methyl]-4-(trifluormethyl)benzolchlorid
1-[(Ammoniooxy)methyl]-4-(trifluormethyl)benzolchlorid
Formel:
C₈H₉ClF₃NO Molecular Weight: 227.61 g/mol |
MDL:
MFCD00231881 CAS-Nummer: 321574-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...