1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
Lieferant: BIONET RESEARCH
Synonyme:
O-(4-Fluorobenzyl)hydroxylamine hydrochloride
3F-940.500
3F-940.1
3F-940.5
3F-940.10
BIOT3F-940.500EA
0
EUR
BIOT3F-940.500
BIOT3F-940.1
BIOT3F-940.5
BIOT3F-940.10
1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
Formel:
C₇H₉ClFNO Molecular Weight: 177,6039 g/mol |
MDL:
MFCD01114581 CAS-Nummer: 51572-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...