1-[(Ammoniooxy)methyl]-2,3-dichlorbenzolchlorid
Lieferant: BIONET RESEARCH
Synonyme:
1-[(Ammoniooxy)methyl]-2,3-dichlorobenzene chloride
1G-926.500
1G-926.1
1G-926.5
1G-926.10
BIOT1G-926.500EA
0
EUR
BIOT1G-926.500
BIOT1G-926.1
BIOT1G-926.5
BIOT1G-926.10
1-[(Ammoniooxy)methyl]-2,3-dichlorbenzolchlorid
1-[(Ammoniooxy)methyl]-2,3-dichlorbenzolchlorid
Formel:
C₇H₇Cl₂NO·HCl Molecular Weight: 228.504 g/mol |
MDL:
MFCD01114575 CAS-Nummer: 317821-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...