O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride

Lieferant: BIONET RESEARCH

Synonyme: 2-[(Ammoniooxy)methyl]-1,4-dichlorobenzene chloride

1G-927.500 1G-927.1 1G-927.5 1G-927.10
BIOT1G-927.500EA 0 EUR
BIOT1G-927.500 BIOT1G-927.1 BIOT1G-927.5 BIOT1G-927.10
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride
Formel: C₇H₇Cl₂NO·HCl
Molecular Weight: 228.504 g/mol
MDL: MFCD01114576
CAS-Nummer: 317821-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR