1-[(Ammoniooxy)methyl]-2-chlorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(2-Chlorobenzyl)hydroxylamine hydrochloride

8G-933.500 8G-933.1 8G-933.5 8G-933.10
BIOT8G-933.500EA 0 EUR
BIOT8G-933.500 BIOT8G-933.1 BIOT8G-933.5 BIOT8G-933.10
1-[(Ammoniooxy)methyl]-2-chlorbenzolchlorid
1-[(Ammoniooxy)methyl]-2-chlorbenzolchlorid
Formel: C₇H₉Cl₂NO
Molecular Weight: 194.0585 g/mol
MDL: MFCD01114592
CAS-Nummer: 5555-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR