4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid
Lieferant: BIONET RESEARCH
Synonyme:
O-[(3,4-Dichlorophenyl)methyl]hydroxylamine hydrochloride, O-(3,4-Dichlorobenzyl)hydroxylamine hydrochloride
5E-042.500
5E-042.1
5E-042.5
5E-042.10
BIOT5E-042.500EA
0
EUR
BIOT5E-042.500
BIOT5E-042.1
BIOT5E-042.5
BIOT5E-042.10
4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid
4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid
Formel:
C₇H₈Cl₃NO Molecular Weight: 228.5035 g/mol |
MDL:
MFCD01114587 CAS-Nummer: 15256-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...