1-[(Ammoniooxy)methyl]-3-chlorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(3-Chlorobenzyl)hydroxylamine hydrochloride

10F-930.500 10F-930.1 10F-930.5 10F-930.10
BIOT10F-930.500EA 0 EUR
BIOT10F-930.500 BIOT10F-930.1 BIOT10F-930.5 BIOT10F-930.10
1-[(Ammoniooxy)methyl]-3-chlorbenzolchlorid
1-[(Ammoniooxy)methyl]-3-chlorbenzolchlorid
Formel: C₇H₉Cl₂NO
MDL: MFCD01114569
CAS-Nummer: 29605-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR