1-[(Ammoniooxy)methyl]-4-chlorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(4-Chlorobenzyl)hydroxylamine hydrochloride

5E-044.500 5E-044.1 5E-044.5 5E-044.10
BIOT5E-044.500EA 0 EUR
BIOT5E-044.500 BIOT5E-044.1 BIOT5E-044.5 BIOT5E-044.10
1-[(Ammoniooxy)methyl]-4-chlorbenzolchlorid
1-[(Ammoniooxy)methyl]-4-chlorbenzolchlorid
Formel: C₇H₉Cl₂NO
Molecular Weight: 194,0585 g/mol
MDL: MFCD01114589
CAS-Nummer: 38936-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR