1-[(Ammoniooxy)methyl]-4-methylbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-[(4-Methylphenyl)methyl]hydroxylamine hydrochloride

12E-920.500 12E-920.1 12E-920.5 12E-920.10
BIOT12E-920.500EA 0 EUR
BIOT12E-920.500 BIOT12E-920.1 BIOT12E-920.5 BIOT12E-920.10
1-[(Ammoniooxy)methyl]-4-methylbenzolchlorid
1-[(Ammoniooxy)methyl]-4-methylbenzolchlorid
Formel: C₈H₁₂ClNO
MDL: MFCD01114570
CAS-Nummer: 38936-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR