1-(2-(Ammoniooxy)ethoxy)-3-(trifluormethyl)benzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-{2-[3-(Trifluoromethyl)phenoxy]ethyl}hydroxylamine hydrochloride

4F-958.500 4F-958.1 4F-958.5 4F-958.10
BIOT4F-958.500EA 0 EUR
BIOT4F-958.500 BIOT4F-958.1 BIOT4F-958.5 BIOT4F-958.10
1-(2-(Ammoniooxy)ethoxy)-3-(trifluormethyl)benzolchlorid
1-(2-(Ammoniooxy)ethoxy)-3-(trifluormethyl)benzolchlorid
Formel: C₉H₁₁ClF₃NO₂
MDL: MFCD01114594
CAS-Nummer: 113211-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR