1-(2-(Ammoniooxy)ethoxy)benzolchlorid

Lieferant: BIONET RESEARCH

3F-936.500 3F-936.1 3F-936.5 3F-936.10
BIOT3F-936.500EA 0 EUR
BIOT3F-936.500 BIOT3F-936.1 BIOT3F-936.5 BIOT3F-936.10
1-(2-(Ammoniooxy)ethoxy)benzolchlorid
1-(2-(Ammoniooxy)ethoxy)benzolchlorid
Formel: C₈H₁₂ClNO₂
MDL: MFCD01114593
CAS-Nummer: 5397-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR