1-[(Ammoniooxy)methyl]-2-methoxybenzolchlorid
Lieferant: BIONET RESEARCH
Synonyme:
O-[(2-Methoxyphenyl)methyl]hydroxylamine hydrochloride
1G-939.500
1G-939.1
1G-939.5
1G-939.10
BIOT1G-939.500EA
0
EUR
BIOT1G-939.500
BIOT1G-939.1
BIOT1G-939.5
BIOT1G-939.10
1-[(Ammoniooxy)methyl]-2-methoxybenzolchlorid
1-[(Ammoniooxy)methyl]-2-methoxybenzolchlorid
Formel:
C₈H₁₂ClNO₂ |
MDL:
MFCD01114579 CAS-Nummer: 317821-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...