1-[(Ammoniooxy)methyl]-3-methoxybenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(3-Methoxybenzyl)hydroxylamine hydrochloride

4F-922.500 4F-922.1 4F-922.5 4F-922.10
BIOT4F-922.500EA 0 EUR
BIOT4F-922.500 BIOT4F-922.1 BIOT4F-922.5 BIOT4F-922.10
1-[(Ammoniooxy)methyl]-3-methoxybenzolchlorid
1-[(Ammoniooxy)methyl]-3-methoxybenzolchlorid
Formel: C₈H₁₂ClNO₂
Molecular Weight: 189.6394 g/mol
MDL: MFCD01114583
CAS-Nummer: 3839-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR