1-[(Ammoniooxy)methyl]-4-methoxybenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(4-Methoxybenzyl)hydroxylamine hydrochloride

4F-903.500 4F-903.1 4F-903.5 4F-903.10
BIOT4F-903.500EA 0 EUR
BIOT4F-903.500 BIOT4F-903.1 BIOT4F-903.5 BIOT4F-903.10
1-[(Ammoniooxy)methyl]-4-methoxybenzolchlorid
1-[(Ammoniooxy)methyl]-4-methoxybenzolchlorid
Formel: C₈H₁₂ClNO₂
Molecular Weight: 189.64 g/mol
MDL: MFCD01114582
CAS-Nummer: 876-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR