D(+)-Allose, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, D-(+)-Allose, β-D-Allopyranose β-D-Allose All, D-Allose

M-Clarity™ Qualitätsstufe = MQ100

285005-100MG 285005-250MG
SIAL285005-100MGEA 58.3 EUR
SIAL285005-100MG SIAL285005-250MG
D(+)-Allose, Sigma-Aldrich®
D(+)-Allose
Formel: C₆H₁₂O₆
Molecular Weight: 180.1559 g/mol
MDL: MFCD00135833
CAS-Nummer: 2595-97-3
EINECS: 219-994-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR