3-(4-Chlorphenyl)-1H-pyrazol-5-amin

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-5-(4-chlorophenyl)pyrazole, 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine

3Y-0844.5 3Y-0844.10 3Y-0844.500 3Y-0844.1
BIOT3Y-0844.5EA 0 EUR
BIOT3Y-0844.5 BIOT3Y-0844.10 BIOT3Y-0844.500 BIOT3Y-0844.1
3-(4-Chlorphenyl)-1H-pyrazol-5-amin
3-(4-Chlorphenyl)-1H-pyrazol-5-amin
Formel: C₉H₈ClN₃
Molecular Weight: 193,64 g/mol
MDL: MFCD00053046
CAS-Nummer: 78583-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR