3-(4-Chlorphenyl)-1H-pyrazol-5-amin
Lieferant: FLUOROCHEM
Synonyme:
3-Amino-5-(4-chlorophenyl)pyrazole, 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine
019533-5G
019533-1G
FLUO019533-5GEA
288
EUR
FLUO019533-5G
FLUO019533-1G
3-(4-Chlorphenyl)-1H-pyrazol-5-amin
3-(4-Chlorphenyl)-1H-pyrazol-5-amin
Formel:
C₉H₈ClN₃ Molecular Weight: 193,64 g/mol |
MDL:
MFCD00053046 CAS-Nummer: 78583-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...