3-(4-Chlorphenyl)-1H-pyrazol-5-amin

Lieferant: Thermo Scientific

Synonyme: 3-Amino-5-(4-chlorophenyl)pyrazole, 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine

KM01548DA KM01548EA
MAYBKM01548DAEA 0 EUR
MAYBKM01548DA MAYBKM01548EA
3-(4-Chlorphenyl)-1H-pyrazol-5-amin
3-(4-Chlorphenyl)-1H-pyrazol-5-amin
Formel: C₉H₈ClN₃
Molecular Weight: 193,64 g/mol
MDL: MFCD00053046
CAS-Nummer: 78583-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR