5-Amino-4-cyan-1-phenyl-1H-pyrazol

Lieferant: FLUOROCHEM

Synonyme: 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

005981-1G 005981-5G
FLUO005981-1GEA 42.9 EUR
FLUO005981-1G FLUO005981-5G
5-Amino-4-cyan-1-phenyl-1H-pyrazol
5-Amino-4-cyano-1-phenyl-1H-pyrazole
Formel: C₁₀H₈N₄
Molecular Weight: 184.20 g/mol
MDL: MFCD00020730
CAS-Nummer: 5334-43-0
EINECS: 226-253-9
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR