3-(4-Fluorphenyl)-1H-pyrazol-5-amin
Lieferant: FLUOROCHEM
Synonyme:
5-Amino-3-(4-fluorophenyl)-1H-pyrazole
019534-2.5G
019534-5G
FLUO019534-2.5GEA
0
EUR
FLUO019534-2.5G
FLUO019534-5G
3-(4-Fluorphenyl)-1H-pyrazol-5-amin
3-(4-Fluorphenyl)-1H-pyrazol-5-amin
Formel:
C₉H₈FN₃ |
MDL:
MFCD01023677 CAS-Nummer: 72411-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...