5-Amino-3-(4-methoxyphenyl)-1H-pyrazole 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Amino-3-(4-methoxyphenyl)pyrazole, 3-(4-Methoxyphenyl)-1H-pyrazol-5-amine

H31580.03 H31580.06
H31580.03EA 27.5 EUR
H31580.03 H31580.06
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole 97%
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole

Purity: 97% CAS No.: 19541-95-8 Molecular Formula: C10H11N3O Formula Weight: 189.22 Melting Point: 141-143° MDL No.: MFCD00462192

Formel: C₁₀H₁₁N₃O
Molecular Weight: 189,22 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00462192
CAS-Nummer: 19541-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR