5-Amino-3-(4-methoxyphenyl)-1H-pyrazole ≥97%

Lieferant: CHEMPUR

Synonyme: 5-Amino-3-(4-methoxyphenyl)pyrazole, 3-(4-Methoxyphenyl)-1H-pyrazol-5-amine

FL12259.POR 012259-1G
CHMPFL12259.POREA 0 EUR
CHMPFL12259.POR FLUO012259-1G
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole ≥97%
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189,22 g/mol
MDL: MFCD00462192
CAS-Nummer: 19541-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR