Diethyl-aminomalonat Hydrochlorid
Lieferant: Thermo Scientific
Synonyme: Diethyl 2-aminomalonate hydrochloride
SB00700.1 SB00700.10 SB00700.50
MAYBSB00700.1EA 0 EUR
MAYBSB00700.1 MAYBSB00700.10 MAYBSB00700.50
Diethyl-aminomalonat Hydrochlorid
Diethyl-aminomalonat Hydrochlorid
Formel: C₇H₁₄ClNO₄
Molecular Weight: 211.65 g/mol
MDL: MFCD00012510
CAS-Nummer: 13433-00-6
EINECS: 236-556-8