5-Amino-4-cyan-3-methyl-1-(3-nitrophenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1427-5G HI-1427-1G
COBBHI-1427-5GEA 595 EUR
COBBHI-1427-5G COBBHI-1427-1G
5-Amino-4-cyan-3-methyl-1-(3-nitrophenyl)-1H-pyrazol
5-Amino-4-cyano-3-methyl-1-(3-nitrophenyl)-1H-pyrazole
Formel: C₁₁H₉N₅O₂
MDL: MFCD11053855
CAS-Nummer: 107842-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR