5-Amino-4-cyan-3-methyl-1-(4-nitrophenyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1397-5G HI-1397-1G
COBBHI-1397-5GEA 451 EUR
COBBHI-1397-5G COBBHI-1397-1G
5-Amino-4-cyan-3-methyl-1-(4-nitrophenyl)-1H-pyrazol
5-Amino-4-cyano-3-methyl-1-(4-nitrophenyl)-1H-pyrazole
Formel: C₁₁H₉N₅O₂
MDL: MFCD10699682
CAS-Nummer: 76982-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR