5-Amino-4-cyan-1-(2-fluorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1418-5G HI-1418-1G
COBBHI-1418-5GEA 451 EUR
COBBHI-1418-5G COBBHI-1418-1G
5-Amino-4-cyan-1-(2-fluorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(2-fluorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₁H₉FN₄
MDL: MFCD10699687
CAS-Nummer: 1072944-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR