5-Amino-4-cyan-1-(4-fluorphenyl)-3-methyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

HI-1407-1G HI-1407-5G
COBBHI-1407-1GEA 137 EUR
COBBHI-1407-1G COBBHI-1407-5G
5-Amino-4-cyan-1-(4-fluorphenyl)-3-methyl-1H-pyrazol
5-Amino-4-cyano-1-(4-fluorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₁H₉FN₄
MDL: MFCD08459267
CAS-Nummer: 51516-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR