5-Amino-1-methyl-3-phenyl-1H-pyrazol
Lieferant: FLUOROCHEM
Synonyme:
1-Methyl-3-phenyl-1H-pyrazol-5-amine
CFF400.10
CFF400.25
017423-10G
017423-25G
017423-1G
017423-5G
CHMPCFF400.10EA
532
EUR
CHMPCFF400.10
CHMPCFF400.25
FLUO017423-10G
FLUO017423-25G
FLUO017423-1G
FLUO017423-5G
5-Amino-1-methyl-3-phenyl-1H-pyrazol
5-Amino-1-methyl-3-phenyl-1H-pyrazole
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173.22 g/mol |
MDL:
MFCD00067874 CAS-Nummer: 10199-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...