5-Amino-3-methyl-1-phenyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methyl-1-phenyl-1H-pyrazol-5-amine, 5-Amino-3-methyl-1-phenylpyrazole, 3-methyl-1-phenyl-1H-pyrazol-5-amine

M-Clarity™ Qualitätsstufe = MQ100

541001-25G 541001-5G
SIAL541001-25GEA 113 EUR
SIAL541001-25G SIAL541001-5G
5-Amino-3-methyl-1-phenyl-1H-pyrazol, Sigma-Aldrich®
5-Amino-3-methyl-1-phenyl-1H-pyrazole
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,21 g/mol
MDL: MFCD00020727
CAS-Nummer: 1131-18-6
EINECS: 214-463-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR