Chlorhexidin zur Synthese

Lieferant: Merck
Gefahr

Synonyme: 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]

M-Clarity™ Qualitätsstufe = MQ200

8.43969.0050 8.43969.0250
8.43969.0050EA 81.4 EUR
8.43969.0050 8.43969.0250
Chlorhexidin zur Synthese
Chlorhexidin
Formel: C₂₂H₃₀Cl₂N₁₀
Molecular Weight: 505,4466 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009673
CAS-Nummer: 55-56-1
EINECS: 200-238-7
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Hill Formula C₂₂H₃₀Cl₂N₁₀
Molar Mass 505.45 g/mol
Melting Point 134 - 136 °C
Solubility 0.8 g/l (20 °C)
LD 50 oral LD50 Rat 5000 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering
Categories of danger irritant, dangerous for the environment
R Phrase R 41-51/53
S Phrase S 26-39-61

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR