Chlorhexidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]

M-Clarity™ Qualitätsstufe = MQ100

282227-1G 282227-5G
SIAL282227-1GEA 133 EUR
SIAL282227-1G SIAL282227-5G
Chlorhexidin, Sigma-Aldrich®
Chlorhexidin
Formel: C₂₂H₃₀Cl₂N₁₀
Molecular Weight: 505,4466 g/mol
MDL: MFCD00009673
CAS-Nummer: 55-56-1
EINECS: 200-238-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR