N-(2-Hydroxyethyl)-2-phenylacetamid

Lieferant: BIONET RESEARCH

8F-926.500 8F-926.1 8F-926.5 8F-926.10
BIOT8F-926.500EA 0 EUR
BIOT8F-926.500 BIOT8F-926.1 BIOT8F-926.5 BIOT8F-926.10
N-(2-Hydroxyethyl)-2-phenylacetamid
N-(2-Hydroxyethyl)-2-phenylacetamid
Formel: C₁₀H₁₃NO₂
MDL: MFCD00043506
CAS-Nummer: 6269-99-4
EINECS: 228-444-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR